Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTSB SIRNA Assays ›
Gene Symbol:
CTSB
Gene Name:
cathepsin B
Gene Aliases:
APPS, CPSB
Chromosome Location:
Chr.8: 11842524 - 11868150 on Build GRCh38
UniGene ID:
Hs.520898
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4087
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTSB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317237.1
NP_001304166.1
7, 8
815
NM_001908.4
NP_001899.1
7, 8
846
NM_147780.3
NP_680090.1
9, 10
1008
NM_147781.3
NP_680091.1
8, 9
965
NM_147782.3
NP_680092.1
8, 9
934
NM_147783.3
NP_680093.1
8, 9
920
XM_006716244.2
XP_006716307.1
9, 10
1032
XM_006716245.2
XP_006716308.1
7, 8
1136
XM_011543812.2
XP_011542114.1
9, 10
941
XM_017013097.1
XP_016868586.1
8, 9
922
XM_017013098.1
XP_016868587.1
8, 9
922
XM_017013099.1
XP_016868588.1
9, 10
964
XM_017013100.1
XP_016868589.1
8, 9
866
XM_017013101.1
XP_016868590.1
4, 5
389
GenBank mRNA
AK075393.1
-
7, 8
822
AK092070.1
-
8, 9
857
AK097384.1
-
2, 3
776
AK130184.1
-
7, 8
783
AK290239.1
-
7, 8
783
AK296845.1
-
6, 7
587
AK297687.1
-
5, 6
547
BC010240.1
-
8, 9
863
4154307
BC095408.1
-
7, 8
783
30334082
BX647765.1
-
7, 8
706
BX647952.1
-
7, 8
786
BX648239.1
-
4, 5
4857
L16510.1
-
8, 9
848
M13230.1
-
3, 4
281
M14221.1
-
8, 9
865
Y18460.1
-
3, 4
353
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
toll-like receptor signaling pathway
proteolysis
autophagy
spermatogenesis
skeletal muscle tissue development
response to wounding
response to glucose
response to organic cyclic compound
response to amine
collagen catabolic process
epithelial cell differentiation
regulation of apoptotic process
response to peptide hormone
response to ethanol
decidualization
viral entry into host cell
regulation of catalytic activity
proteolysis involved in cellular protein catabolic process
negative regulation of cell death
response to interleukin-4
cellular response to mechanical stimulus
cellular response to thyroid hormone stimulus
cysteine-type endopeptidase activity
serine-type endopeptidase activity
protein binding
collagen binding
peptidase activity
cysteine-type peptidase activity
kininogen binding
peptide binding
proteoglycan binding
protein self-association
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0