Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284265.1 | NP_001271194.1 | 2, 3 | 138 | ||
NM_173510.3 | NP_775781.1 | 2, 3 | 412 | |||
GenBank mRNA | AK091133.1 | - | 2, 3 | 338 | ||
AK289516.1 | - | 2, 3 | 410 | |||
AK302798.1 | - | 2, 3 | 141 | |||
BC040488.1 | - | 2, 3 | 367 | 5272169 | ||
BC053874.1 | - | 2, 3 | 372 | 5531689 | ||
BC065549.1 | - | 2, 3 | 374 | 6045091 | ||
DB087275.1 | - | 2, 3 | 139 |