Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284263.1 | NP_001271192.1 | 2, 3 | 594 | ||
NM_001284265.1 | NP_001271194.1 | 3, 4 | 374 | |||
NM_173510.3 | NP_775781.1 | 3, 4 | 648 | |||
GenBank mRNA | AK091133.1 | - | 3, 4 | 574 | ||
AK289516.1 | - | 3, 4 | 646 | |||
AK302798.1 | - | 3, 4 | 377 | |||
AK302925.1 | - | 2, 3 | 540 | |||
BC040488.1 | - | 3, 4 | 603 | 5272169 | ||
BC053874.1 | - | 3, 4 | 608 | 5531689 | ||
BC065549.1 | - | 3, 4 | 610 | 6045091 | ||
CR456461.1 | - | 2, 3 | 171 | |||
DB087275.1 | - | 3, 4 | 376 |