Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136505.1 | NP_001129977.1 | 7 | 700 | ||
XM_011523004.2 | XP_011521306.1 | 7 | 1778 | |||
XM_011523005.2 | XP_011521307.1 | 7 | 736 | |||
XM_011523006.2 | XP_011521308.1 | 7 | 1877 | |||
XM_011523007.2 | XP_011521309.1 | 6 | 1804 | |||
XM_011523008.2 | XP_011521310.1 | 6 | 570 | |||
XM_011523009.2 | XP_011521311.1 | 6 | 511 | |||
XM_011523010.2 | XP_011521312.1 | 6 | 1818 | |||
XM_011523011.2 | XP_011521313.1 | 5 | 1741 | |||
XM_011523012.2 | XP_011521314.1 | 5 | 515 | |||
XM_011523014.2 | XP_011521316.1 | 7 | 700 | |||
XM_011523015.2 | XP_011521317.1 | 7 | 700 | |||
XM_011523016.2 | XP_011521318.1 | 7 | 700 | |||
XM_011523017.1 | XP_011521319.1 | 7 | 700 | |||
XM_011523018.1 | XP_011521320.1 | 7 | 700 | |||
XM_011523020.1 | XP_011521322.1 | 7 | 700 | |||
XM_017023157.1 | XP_016878646.1 | 7 | 700 | |||
XM_017023158.1 | XP_016878647.1 | 7 | 700 | |||
XM_017023159.1 | XP_016878648.1 | 7 | 700 | |||
GenBank mRNA | AK093213.1 | - | 7 | 702 | ||
BC126109.1 | - | 7 | 669 | 8991825 |