Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173628.3 | NP_775899.3 | 8 | 1223 | ||
XM_011525416.2 | XP_011523718.1 | 8 | 1259 | |||
GenBank mRNA | AK097776.1 | - | 4 | 310 | ||
BC105107.1 | - | 3 | 267 | |||
BC112284.1 | - | 3 | 267 |