Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174932.2 | NP_777592.1 | 1 | 101 | ||
XM_011530088.2 | XP_011528390.1 | 3 | 281 | |||
XM_011530089.1 | XP_011528391.1 | 3 | 238 | |||
XM_011530090.1 | XP_011528392.1 | 2 | 139 | |||
XM_011530091.2 | XP_011528393.1 | 3 | 281 | |||
XM_017028740.1 | XP_016884229.1 | 3 | 281 | |||
GenBank mRNA | AF465766.1 | - | 1 | 101 | ||
BC131582.1 | - | 1 | 101 | 40112599 |