Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1PR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320730.1 | NP_001307659.1 | 2 | 2437 | ||
NM_001400.4 | NP_001391.2 | 2 | 2545 | |||
GenBank mRNA | BC018650.1 | - | 2 | 2279 | 4071217 | |
M31210.1 | - | 2 | 2278 |
Set Membership: |
![]() |