Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173825.3 | NP_776186.2 | 6 | 603 | ||
XM_017006227.1 | XP_016861716.1 | 6 | 587 | |||
GenBank mRNA | AK025772.1 | - | 3 | 299 | ||
AK293085.1 | - | 6 | 587 | |||
BC020832.1 | - | 6 | 582 | 4771781 | ||
BX647559.1 | - | 6 | 600 | |||
BX647593.1 | - | 6 | 603 | |||
BX647664.1 | - | 6 | 587 |