Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319053.1 | NP_001305982.1 | 19 | 2462 | ||
NM_001470.3 | NP_001461.1 | 19 | 2526 | |||
NM_021903.2 | NP_068703.1 | 14 | 2326 | |||
NM_021904.3 | NP_068704.2 | 18 | 2340 | |||
XM_005248982.2 | XP_005249039.1 | 19 | 2341 | |||
XM_006715047.3 | XP_006715110.1 | 18 | 2141 | |||
XM_011514453.2 | XP_011512755.1 | 17 | 2120 | |||
XM_017010676.1 | XP_016866165.1 | 15 | 1872 | |||
GenBank mRNA | AB208790.1 | - | 19 | 2261 | ||
AF099148.1 | - | 19 | 2348 | |||
AJ012185.1 | - | 18 | 2240 | |||
AJ012186.1 | - | 14 | 1889 | |||
AJ012187.1 | - | 17 | 2054 | |||
AJ012288.1 | - | 19 | 2283 | |||
AJ225028.1 | - | 19 | 2474 | |||
AJ225029.1 | - | 14 | 2068 | |||
AK223619.1 | - | 14 | 2111 | |||
AK290191.1 | - | 14 | 2111 | |||
AK312308.1 | - | 14 | 2107 | |||
BC023336.1 | - | 4 | 384 | |||
BC041332.2 | - | 14 | 2111 | 5275467 | ||
BC042598.1 | - | 14 | 2100 | 4938491 | ||
BC050532.1 | - | 14 | 2098 | 5732186 | ||
Y11044.1 | - | 18 | 2237 |
Set Membership: |
![]() |