Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41510
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
4
390
NM_001195249.1
NP_001182178.1
4
559
NM_001195250.1
NP_001182179.1
3
228
NM_001195251.1
NP_001182180.1
5
496
NM_001195254.1
NP_001182183.1
3
250
NM_175069.2
NP_778239.1
4
390
NM_175073.2
NP_778243.1
5
496
NR_036576.1
4
385
NR_036577.1
4
275
NR_036578.1
4
407
NR_036579.1
4
554
XM_006716791.3
XP_006716854.1
4
398
XM_006716792.2
XP_006716855.1
4
416
XM_011517938.1
XP_011516240.1
3
374
XM_011517939.2
XP_011516241.1
3
329
XM_017014831.1
XP_016870320.1
4
390
XM_017014832.1
XP_016870321.1
4
390
XM_017014833.1
XP_016870322.1
4
390
XM_017014834.1
XP_016870323.1
4
390
XM_017014835.1
XP_016870324.1
4
390
XM_017014836.1
XP_016870325.1
4
390
XM_017014837.1
XP_016870326.1
3
479
XM_017014838.1
XP_016870327.1
4
372
GenBank mRNA
AB603741.1
-
3
286
AJ565850.1
-
4
248
AJ565851.1
-
4
253
AJ565852.1
-
4
363
AJ565853.1
-
3
366
AJ565854.1
-
3
371
AJ565855.1
-
2
209
AJ575566.1
-
3
251
AK000164.1
-
3
393
AK055672.1
-
4
377
AK131046.1
-
4
342
AY040777.1
-
4
365
AY208829.1
-
4
365
AY208830.1
-
3
228
AY208832.1
-
3
203
AY208833.1
-
3
203
AY208834.1
-
4
361
AY208835.1
-
4
345
AY208836.1
-
4
505
AY208837.1
-
5
462
AY208838.1
-
3
321
AY208839.1
-
4
499
AY208840.1
-
4
362
AY208841.1
-
3
199
AY302067.1
-
3
508
AY302068.1
-
3
503
AY302069.1
-
3
393
AY302070.1
-
3
388
AY302071.1
-
3
508
AY302072.1
-
3
503
AY302073.1
-
3
393
AY302074.1
-
3
388
BC001628.1
-
3
213
BC032650.1
-
4
431
5505120
BC068107.1
-
5
471
BC104881.1
-
4
404
8143884
BQ957536.1
-
5
479
BX538161.1
-
5
475
DA664623.1
-
3
417
DB042208.1
-
3
250
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0