Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256113.1 | NP_001243042.1 | 2 | 226 | ||
NM_175075.4 | NP_778250.2 | 3 | 394 | |||
XM_006716232.3 | XP_006716295.1 | 2 | 374 | |||
XM_017013113.1 | XP_016868602.1 | 2 | 708 | |||
GenBank mRNA | AY194292.2 | - | 2 | 212 | ||
BC136248.1 | - | 2 | 373 | 9020235 | ||
BC136249.1 | - | 2 | 373 | 9020236 | ||
CN430635.1 | - | 2 | 205 | |||
DB461052.2 | - | 3 | 388 | |||
FJ381685.1 | - | 2 | 218 |