Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174978.2 | NP_777638.2 | 5 | 400 | ||
XM_011536690.2 | XP_011534992.1 | 6 | 530 | |||
XM_011536703.2 | XP_011535005.1 | 6 | 530 | |||
XM_011536704.1 | XP_011535006.1 | 6 | 531 | |||
XM_017021247.1 | XP_016876736.1 | 6 | 531 | |||
XM_017021248.1 | XP_016876737.1 | 4 | 363 | |||
XM_017021249.1 | XP_016876738.1 | 2 | 95 | |||
XM_017021251.1 | XP_016876740.1 | 1 | 41 | |||
GenBank mRNA | AK098187.1 | - | 5 | 385 | ||
AL832219.1 | - | 5 | 450 | |||
BC125070.1 | - | 5 | 371 | 40117603 | ||
DB447488.2 | - | 5 | 419 |