Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41588
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
6
786
NM_001195249.1
NP_001182178.1
6
955
NM_001195250.1
NP_001182179.1
5
624
NM_001195251.1
NP_001182180.1
7
892
NM_001195252.1
NP_001182181.1
6
570
NM_001195254.1
NP_001182183.1
5
646
NM_175069.2
NP_778239.1
6
786
NM_175073.2
NP_778243.1
7
892
NR_036576.1
6
781
NR_036577.1
6
671
NR_036578.1
6
803
NR_036579.1
6
950
XM_006716791.3
XP_006716854.1
6
794
XM_006716792.2
XP_006716855.1
6
812
XM_011517938.1
XP_011516240.1
5
770
XM_011517939.2
XP_011516241.1
5
725
XM_017014831.1
XP_016870320.1
6
786
XM_017014832.1
XP_016870321.1
6
786
XM_017014833.1
XP_016870322.1
6
786
XM_017014834.1
XP_016870323.1
6
786
XM_017014835.1
XP_016870324.1
6
786
XM_017014836.1
XP_016870325.1
6
786
XM_017014837.1
XP_016870326.1
5
875
XM_017014838.1
XP_016870327.1
6
768
GenBank mRNA
AB603741.1
-
5
682
AJ565850.1
-
6
644
AJ565851.1
-
6
649
AJ565852.1
-
6
759
AJ565853.1
-
5
762
AJ565854.1
-
5
767
AJ565855.1
-
4
605
AJ575566.1
-
5
647
AK000164.1
-
5
789
AK055672.1
-
6
773
AK131046.1
-
6
738
AY040777.1
-
6
761
AY208829.1
-
6
761
AY208830.1
-
5
624
AY208831.1
-
6
536
AY208832.1
-
5
599
AY208833.1
-
5
599
AY208834.1
-
5
700
AY208835.1
-
6
741
AY208836.1
-
6
901
AY208837.1
-
7
858
AY208838.1
-
5
717
AY208839.1
-
6
895
AY208840.1
-
6
758
AY208841.1
-
5
595
AY208842.1
-
3
395
AY302067.1
-
5
904
AY302068.1
-
5
899
AY302069.1
-
5
789
AY302070.1
-
5
784
BC001628.1
-
5
609
BC032650.1
-
6
827
5505120
BC068107.1
-
7
867
BC104881.1
-
6
800
8143884
BX538161.1
-
7
871
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-xm8cg:80/10.239.167.213:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0