Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNTN4 SIRNA Assays ›
Gene Symbol:
CNTN4
Gene Name:
contactin 4
Gene Aliases:
AXCAM, BIG-2
Chromosome Location:
Chr.3: 2098771 - 3059080 on Build GRCh38
UniGene ID:
Hs.298705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41594
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CNTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001206955.1
NP_001193884.1
20, 21
2903
NM_001206956.1
NP_001193885.1
12, 13
1622
NM_175607.2
NP_783200.1
21, 22
2858
NM_175613.2
NP_783302.1
12, 13
1625
XM_006713004.3
XP_006713067.1
20, 21
2868
XM_011533425.2
XP_011531727.1
20, 21
2871
XM_011533426.2
XP_011531728.1
21, 22
3090
XM_011533427.2
XP_011531729.1
21, 22
3071
XM_011533428.2
XP_011531730.1
19, 20
3060
XM_011533429.2
XP_011531731.1
19, 20
2995
XM_011533430.2
XP_011531732.1
19, 20
2956
XM_011533431.2
XP_011531733.1
18, 19
5705
XM_017005782.1
XP_016861271.1
21, 22
2813
XM_017005783.1
XP_016861272.1
22, 23
3687
XM_017005784.1
XP_016861273.1
20, 21
3148
XM_017005785.1
XP_016861274.1
21, 22
2795
XM_017005786.1
XP_016861275.1
19, 20
3057
XM_017005787.1
XP_016861276.1
19, 20
2953
GenBank mRNA
AF464063.1
-
2, 3
769
AF549455.1
-
18, 19
2901
AK057278.1
-
12, 13
1603
AK097633.1
-
15, 16
2149
AK125460.1
-
20, 21
3046
AK314396.1
-
12, 13
1603
AY090737.1
-
20, 21
2740
BC026119.1
-
12, 13
1605
4826091
Back To Top
More Information
Gene Ontology Categories:
Function(s)
neuron cell-cell adhesion
nervous system development
axonogenesis
axon guidance
axonal fasciculation
brain development
neuron projection development
negative regulation of neuron differentiation
regulation of synaptic plasticity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0