Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198569.1 | NP_001185498.1 | 4 | 571 | ||
NM_001694.3 | NP_001685.1 | 3 | 730 | |||
GenBank mRNA | BC004537.2 | - | 3 | 614 | 3618755 | |
BC007389.2 | - | 3 | 618 | 3831016 | ||
BC007759.2 | - | 3 | 602 | 4127653 | ||
BC009290.2 | - | 3 | 635 | 4111426 | ||
M62762.1 | - | 3 | 726 |