Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278639.1 | NP_001265568.1 | 5, 6 | 978 | ||
NM_002882.3 | NP_002873.1 | 5, 6 | 726 | |||
XM_011530291.2 | XP_011528593.1 | 5, 6 | 1040 | |||
GenBank mRNA | CA489474.1 | - | 5, 6 | 668 | ||
CR456556.1 | - | 5, 6 | 554 | |||
X83617.1 | - | 5, 6 | 602 |