Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306146.1 | NP_001293075.1 | 5 | 835 | ||
NM_002112.3 | NP_002103.2 | 5 | 835 | |||
XM_017022094.1 | XP_016877583.1 | 5 | 599 | |||
XM_017022095.1 | XP_016877584.1 | 5 | 599 | |||
XM_017022096.1 | XP_016877585.1 | 4 | 384 | |||
XM_017022097.1 | XP_016877586.1 | 3 | 682 | |||
XM_017022098.1 | XP_016877587.1 | 2 | 202 | |||
XM_017022099.1 | XP_016877588.1 | 5 | 599 | |||
GenBank mRNA | AK308833.1 | - | 5 | 835 | ||
AK310479.1 | - | 3 | 361 | |||
AK311072.1 | - | 5 | 554 | |||
BC130527.1 | - | 5 | 536 | 40146558 | ||
BC144173.1 | - | 5 | 536 | 9052693 | ||
M60445.1 | - | 5 | 544 | |||
X54297.1 | - | 5 | 595 |