Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004708.3 | NP_004699.1 | 3 | 207 | ||
XM_005259392.4 | XP_005259449.1 | 3 | 234 | |||
GenBank mRNA | AB209040.1 | - | 3 | 2802 | ||
AF014955.1 | - | 3 | 157 | |||
AK124774.1 | - | 1 | 1021 | |||
AK293486.1 | - | 3 | 205 | |||
BC015519.2 | - | 3 | 173 | 3888459 | ||
BM696336.1 | - | 3 | 197 | |||
BT006694.1 | - | 3 | 133 | |||
CR456934.1 | - | 3 | 133 | |||
DN915905.1 | - | 3 | 207 |