Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002801.3 | NP_002792.1 | 4 | 445 | ||
GenBank mRNA | AK225658.1 | - | 4 | 418 | ||
AK298012.1 | - | 2 | 753 | |||
AK312208.1 | - | 4 | 405 | |||
BC052369.1 | - | 4 | 383 | 4054165 | ||
BP289065.1 | - | 4 | 445 | |||
BT019723.1 | - | 4 | 317 | |||
Y13640.1 | - | 4 | 405 |