Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003214.3 | NP_003205.2 | 13 | 2717 | ||
GenBank mRNA | AF142482.1 | - | 13 | 2667 | ||
AK226144.1 | - | 9 | 4077 | |||
BC010053.2 | - | 13 | 1654 | |||
BC027877.1 | - | 11 | 2455 | 5207501 | ||
BC091488.1 | - | 13 | 2642 | 6187474 |