Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005250.2 | NP_005241.1 | 1 | 1000 | ||
GenBank mRNA | BC100027.1 | - | 1 | 1000 | 30920793 | |
BC117226.1 | - | 1 | 828 | 40125777 |