Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPBWR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005286.2 | NP_005277.2 | 1 | 895 | ||
GenBank mRNA | BC067481.1 | - | 1 | 895 | 6971825 | |
BC067482.1 | - | 1 | 895 | 6971826 | ||
BC067483.1 | - | 1 | 895 | 6971827 |
Set Membership: |
![]() |