Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318835.1 | NP_001305764.1 | 4 | 568 | ||
NM_005794.3 | NP_005785.1 | 4 | 856 | |||
NM_182908.4 | NP_878912.1 | 4 | 856 | |||
XM_005267249.1 | XP_005267306.1 | 4 | 568 | |||
XM_006720001.3 | XP_006720064.1 | 4 | 531 | |||
XM_011536338.1 | XP_011534640.1 | 3 | 497 | |||
XM_011536339.2 | XP_011534641.1 | 3 | 347 | |||
GenBank mRNA | AB209653.1 | - | 3 | 1242 | ||
AK222857.1 | - | 4 | 856 | |||
AK309945.1 | - | 4 | 857 | |||
AK309975.1 | - | 4 | 568 | |||
BC002786.2 | - | 4 | 847 | 3614599 | ||
BC007339.2 | - | 3 | 382 | |||
U31875.1 | - | 4 | 842 |