Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006041.2 | NP_006032.1 | 2 | 1801 | ||
XM_017025479.1 | XP_016880968.1 | 2 | 2229 | |||
GenBank mRNA | AF105377.1 | - | 2 | 1662 | ||
BC063301.1 | - | 2 | 1718 | 30343352 | ||
BC069664.1 | - | 2 | 1602 | 7262483 | ||
BC069725.1 | - | 2 | 1601 | 7262495 |