Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014205.3 | NP_055020.1 | 1 | 1078 | ||
GenBank mRNA | AF119497.1 | - | 1 | 1038 | ||
AK027081.1 | - | 1 | 1013 | |||
AK223333.1 | - | 1 | 993 | |||
AK223358.1 | - | 1 | 1048 | |||
BC052240.1 | - | 1 | 1041 | 6107352 |