Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154829.2 | - | 935 | |||
AK000419.1 | - | 935 | ||||
BC008183.1 | - | 952 | 4183256 | |||
BC017454.1 | - | 913 | 4647090 | |||
EU794640.1 | - | 935 |