Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018476.3 | NP_060946.3 | 3 | 431 | ||
GenBank mRNA | AF183416.1 | - | 1 | 193 | ||
AF220189.1 | - | 3 | 357 | |||
AF237783.1 | - | 3 | 362 | |||
AK290958.1 | - | 3 | 365 | |||
AY833561.1 | - | 3 | 353 | |||
BC018615.2 | - | 1 | 197 | |||
BC126427.1 | - | 2 | 241 | 8992143 | ||
BC126429.1 | - | 2 | 241 | 8992145 | ||
BM465948.1 | - | 3 | 372 | |||
BX098763.1 | - | 3 | 431 |