Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143781.1 | NP_001137253.1 | 2 | 121 | ||
NM_001143782.1 | NP_001137254.1 | 3 | 327 | |||
NM_016594.2 | NP_057678.1 | 3 | 327 | |||
GenBank mRNA | AF238079.1 | - | 3 | 280 | ||
AK301455.1 | - | 3 | 300 | |||
AY358998.1 | - | 3 | 285 | |||
BC027973.1 | - | 3 | 279 | 5229643 | ||
BC048292.1 | - | 3 | 290 | 5207367 | ||
DA378205.1 | - | 3 | 327 |