Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023004.5 | NP_075380.1 | 2 | 1662 | ||
GenBank mRNA | AK054602.1 | - | 2 | 1718 | ||
AL834449.1 | - | 2 | 1605 | |||
BC011787.1 | - | 2 | 1507 | 4040540 | ||
CR456360.1 | - | 2 | 1501 |