Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244974.1 | NP_001231903.1 | 2 | 372 | ||
NM_002710.3 | NP_002701.1 | 2 | 372 | |||
XM_011538504.2 | XP_011536806.1 | 2 | 293 | |||
XM_011538505.2 | XP_011536807.1 | 2 | 293 | |||
GenBank mRNA | AK297882.1 | - | 2 | 90 | ||
BC014073.2 | - | 2 | 265 | 3956353 | ||
DA472620.1 | - | 2 | 346 | |||
DA514523.1 | - | 2 | 373 | |||
L07395.1 | - | 2 | 89 | |||
X74008.1 | - | 2 | 255 |