Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP2R5C SIRNA Assays ›
Gene Symbol:
PPP2R5C
Gene Name:
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases:
B56G, PR61G
Chromosome Location:
Chr.14: 101760573 - 101927992 on Build GRCh38
UniGene ID:
Hs.368264
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4260
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP2R5C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161725.1
NP_001155197.1
4
395
NM_001161726.1
NP_001155198.1
4
467
NM_002719.3
NP_002710.2
2
346
NM_178586.2
NP_848701.1
2
346
NM_178587.2
NP_848702.1
2
346
XM_005267819.1
XP_005267876.1
4
456
XM_005267820.1
XP_005267877.1
4
396
XM_005267822.3
XP_005267879.1
4
456
XM_005267823.1
XP_005267880.1
4
456
XM_005267824.1
XP_005267881.1
4
456
XM_005267826.1
XP_005267883.1
2
169
XM_005267827.1
XP_005267884.1
4
391
XM_011536914.2
XP_011535216.1
3
260
XM_011536915.2
XP_011535217.1
3
343
XM_011536916.2
XP_011535218.1
3
1342
XM_011536918.1
XP_011535220.1
3
497
XM_011536919.2
XP_011535221.1
3
479
XM_011536920.1
XP_011535222.1
3
373
XM_011536921.2
XP_011535223.1
2
262
XM_017021422.1
XP_016876911.1
3
412
XM_017021423.1
XP_016876912.1
2
346
XM_017021424.1
XP_016876913.1
3
373
GenBank mRNA
AB451341.1
-
2
206
AI692258.1
-
4
468
AK131391.1
-
4
380
AK302470.1
-
4
295
AK315072.1
-
2
282
BC016183.1
-
2
244
4855950
BI560029.1
-
2
320
D26445.1
-
2
203
DA853338.1
-
2
346
L42375.1
-
2
176
U37352.1
-
2
264
Z69030.1
-
2
284
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
negative regulation of cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
regulation of protein phosphatase type 2A activity
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
protein phosphatase type 2A regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0