Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288710.1 | NP_001275639.1 | 3 | 479 | ||
NM_032366.4 | NP_115742.3 | 4 | 539 | |||
NR_109976.1 | 3 | 483 | ||||
NR_109977.1 | 3 | 376 | ||||
NR_109978.1 | 4 | 646 | ||||
NR_109979.1 | 2 | 316 | ||||
XM_011522713.2 | XP_011521015.1 | 4 | 738 | |||
GenBank mRNA | AK057077.1 | - | 1 | 1437 | ||
AK294482.1 | - | 4 | 475 | |||
AW027232.1 | - | 3 | 395 | |||
BG612714.1 | - | 3 | 340 | |||
BI762987.1 | - | 4 | 515 | |||
BP273946.1 | - | 3 | 415 | |||
BQ921757.1 | - | 444 | ||||
BU539308.1 | - | 2 | 264 |