Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138706.4 | NP_619651.3 | 2 | 1232 | ||
GenBank mRNA | AB073740.1 | - | 1 | 1094 | ||
AK127544.1 | - | 2 | 815 | |||
AK172863.1 | - | 2 | 1157 | |||
AK292773.1 | - | 2 | 1175 | |||
AK313682.1 | - | 2 | 1163 | |||
BC025357.1 | - | 1 | 773 | |||
BC103908.1 | - | 2 | 1131 | 40005856 | ||
BC103909.2 | - | 2 | 1131 | 40005863 | ||
BC103910.1 | - | 2 | 1131 | 40005864 |