Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031275.4 | NP_112565.1 | 5 | 426 | ||
GenBank mRNA | AF285600.1 | - | 4 | 332 | ||
BC029506.1 | - | 5 | 426 | 5298531 | ||
BI561514.1 | - | 5 | 430 |