Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 7, 8 | 864 | ||
NM_001321852.1 | NP_001308781.1 | 6, 7 | 867 | |||
NM_001321853.1 | NP_001308782.1 | 8, 9 | 1126 | |||
NM_001321854.1 | NP_001308783.1 | 7, 8 | 1042 | |||
NM_001321855.1 | NP_001308784.1 | 7, 8 | 1045 | |||
NM_001321856.1 | NP_001308785.1 | 6, 7 | 780 | |||
NM_001321857.1 | NP_001308786.1 | 6, 7 | 961 | |||
NM_002227.3 | NP_002218.2 | 6, 7 | 961 | |||
XM_017001212.1 | XP_016856701.1 | 6, 7 | 808 | |||
GenBank mRNA | AB209057.1 | - | 5, 6 | 717 | ||
AB219242.1 | - | 4, 5 | 611 | |||
AK314257.1 | - | 7, 8 | 866 | |||
BC009079.1 | - | 6, 7 | 896 | |||
BC023604.2 | - | 4, 5 | 652 | |||
BC062431.1 | - | 6, 7 | 896 | |||
BC111401.1 | - | 6, 7 | 926 | |||
BC132729.1 | - | 5, 6 | 679 | 40146751 | ||
HQ628631.1 | - | 4, 5 | 641 | |||
M64174.1 | - | 5, 6 | 686 |