Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021960.4 | NP_068779.1 | 1 | 336 | ||
NM_182763.2 | NP_877495.1 | 1 | 336 | |||
GenBank mRNA | AF118124.1 | - | 1 | 177 | ||
AF203373.1 | - | 1 | 128 | |||
AK297217.1 | - | 1 | 208 | |||
AK312508.1 | - | 1 | 208 | |||
AK316267.1 | - | 2 | 164 | |||
BC017197.2 | - | 1 | 182 | 3138465 | ||
BC071897.1 | - | 1 | 184 | 6586159 | ||
BC107735.1 | - | 1 | 199 | 4770487 | ||
BT006640.1 | - | 1 | 128 | |||
L08246.1 | - | 1 | 188 |