Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287049.1 | NP_001273978.1 | 3 | 400 | ||
NM_153707.3 | NP_714918.2 | 3 | 642 | |||
XM_011517744.1 | XP_011516046.1 | 4 | 722 | |||
XM_011517745.1 | XP_011516047.1 | 5 | 657 | |||
XM_011517746.2 | XP_011516048.1 | 5 | 578 | |||
XM_011517747.2 | XP_011516049.1 | 2 | 238 | |||
XM_017014314.1 | XP_016869803.1 | 5 | 968 | |||
XM_017014315.1 | XP_016869804.1 | 3 | 355 | |||
GenBank mRNA | AK292245.1 | - | 3 | 593 | ||
BC033489.1 | - | 3 | 570 | 5170510 | ||
BM563230.1 | - | 3 | 419 |