Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000156.5 | NP_000147.1 | 4, 5 | 546 | ||
NM_138924.2 | NP_620279.1 | 4, 5 | 546 | |||
GenBank mRNA | AF086508.1 | - | 2, 3 | 131 | ||
AK289465.1 | - | 4, 5 | 520 | |||
AK297487.1 | - | 3, 4 | 405 | |||
BC016760.2 | - | 4, 5 | 493 | 2989341 | ||
BC017936.1 | - | 4, 5 | 491 | 4246521 | ||
BI914772.1 | - | 4, 5 | 529 | |||
BT007034.1 | - | 4, 5 | 452 | |||
EU794612.1 | - | 4, 5 | 452 | |||
Z49878.1 | - | 4, 5 | 495 |