Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 2 | 422 | ||
XM_005249652.3 | XP_005249709.1 | 2 | 365 | |||
GenBank mRNA | AK096533.1 | - | 2 | 365 | ||
AK294982.1 | - | 2 | 152 | |||
AL834391.1 | - | 2 | 356 | |||
AY054121.1 | - | 2 | 218 | |||
BC062423.1 | - | 2 | 173 | |||
BI560774.1 | - | 2 | 426 | |||
DT217816.1 | - | 2 | 357 |