Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037174.1 | NP_001032251.1 | 3, 4 | 324 | ||
NM_012097.3 | NP_036229.1 | 3, 4 | 580 | |||
NM_177985.2 | NP_817114.2 | 3, 4 | 322 | |||
GenBank mRNA | AF087889.1 | - | 510 | |||
AF100740.1 | - | 3, 4 | 327 | |||
AF493891.1 | - | 3, 4 | 261 | |||
AK292714.1 | - | 3, 4 | 490 | |||
AK298937.1 | - | 3, 4 | 268 | |||
BC001254.1 | - | 3, 4 | 498 | 3453177 | ||
BG707385.1 | - | 3, 4 | 301 | |||
BQ216648.1 | - | 3, 4 | 567 | |||
BT009829.1 | - | 3, 4 | 261 | |||
BX461356.2 | - | 3, 4 | 295 | |||
CX867116.1 | - | 3, 4 | 586 |