Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176816.4 | NP_789786.2 | 2, 3 | 412 | ||
XM_005248458.4 | XP_005248515.1 | 2, 3 | 649 | |||
XM_005248464.3 | XP_005248521.1 | 2, 3 | 454 | |||
XM_006714569.3 | XP_006714632.1 | 2, 3 | 454 | |||
XM_011543255.2 | XP_011541557.1 | 2, 3 | 429 | |||
XM_011543256.2 | XP_011541558.1 | 2, 3 | 430 | |||
XM_011543257.2 | XP_011541559.1 | 2, 3 | 454 | |||
XM_011543258.2 | XP_011541560.1 | 2, 3 | 454 | |||
XM_011543259.2 | XP_011541561.1 | 2, 3 | 454 | |||
XM_011543260.2 | XP_011541562.1 | 2, 3 | 454 | |||
XM_017009207.1 | XP_016864696.1 | 2, 3 | 454 | |||
GenBank mRNA | AB024691.1 | - | 2, 3 | 412 |