Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRS2 SIRNA Assays ›
Gene Symbol:
IRS2
Gene Name:
insulin receptor substrate 2
Gene Aliases:
IRS-2
Chromosome Location:
Chr.13: 109753837 - 109786567 on Build GRCh38
UniGene ID:
Hs.442344
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43348
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003749.2
NP_003740.2
1
639
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
positive regulation of mesenchymal cell proliferation
negative regulation of B cell apoptotic process
glucose metabolic process
signal transduction
brain development
cell proliferation
positive regulation of cell proliferation
insulin receptor signaling pathway
response to glucose
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of glucose metabolic process
regulation of phosphatidylinositol 3-kinase signaling
regulation of lipid metabolic process
positive regulation of cell migration
mammary gland development
positive regulation of B cell proliferation
positive regulation of fatty acid beta-oxidation
positive regulation of insulin secretion
cellular response to insulin stimulus
negative regulation of kinase activity
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of GTPase activity
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
lipid homeostasis
cellular response to glucose stimulus
signal transducer activity
Ras guanyl-nucleotide exchange factor activity
insulin receptor binding
protein binding
1-phosphatidylinositol-3-kinase activity
protein kinase binding
protein phosphatase binding
protein domain specific binding
phosphatidylinositol 3-kinase binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0