Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF1 SIRNA Assays ›
Gene Symbol:
IKZF1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
Chromosome Location:
Chr.7: 50303453 - 50405101 on Build GRCh38
UniGene ID:
Hs.435949
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43362
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220765.2
NP_001207694.1
3
403
NM_001220767.2
NP_001207696.1
2
149
NM_001220768.2
NP_001207697.1
2
149
NM_001220770.2
NP_001207699.1
2
149
NM_001220771.2
NP_001207700.1
2
149
NM_001291837.1
NP_001278766.1
3
376
NM_001291838.1
NP_001278767.1
3
376
NM_001291839.1
NP_001278768.1
3
181
NM_001291840.1
NP_001278769.1
2
149
NM_001291841.1
NP_001278770.1
2
149
NM_001291842.1
NP_001278771.1
2
149
NM_001291843.1
NP_001278772.1
2
149
NM_001291844.1
NP_001278773.1
2
149
NM_001291845.1
NP_001278774.1
3
403
NM_001291846.1
NP_001278775.1
3
403
NM_001291847.1
NP_001278776.1
3
376
NM_006060.5
NP_006051.1
3
403
XM_011515058.2
XP_011513360.1
3
233
XM_011515059.2
XP_011513361.1
3
235
XM_011515060.2
XP_011513362.1
3
3383
XM_011515061.2
XP_011513363.1
3
231
XM_011515062.2
XP_011513364.1
3
513
XM_011515063.2
XP_011513365.1
3
430
XM_011515064.2
XP_011513366.1
3
4061
XM_011515065.2
XP_011513367.1
3
198
XM_011515066.2
XP_011513368.1
3
302
XM_011515067.2
XP_011513369.1
3
235
XM_011515068.2
XP_011513370.1
3
430
XM_011515069.2
XP_011513371.1
3
4058
XM_011515070.2
XP_011513372.1
3
234
XM_011515071.2
XP_011513373.1
3
233
XM_011515072.2
XP_011513374.1
3
231
XM_011515073.2
XP_011513375.1
3
513
XM_011515074.2
XP_011513376.1
3
234
XM_011515075.2
XP_011513377.1
3
513
XM_011515076.2
XP_011513378.1
3
457
XM_011515077.2
XP_011513379.1
3
234
XM_011515078.2
XP_011513380.1
3
237
XM_017011667.1
XP_016867156.1
3
430
XM_017011668.1
XP_016867157.1
3
197
XM_017011669.1
XP_016867158.1
3
198
XM_017011670.1
XP_016867159.1
3
559
GenBank mRNA
AF116605.1
-
1
686
AK303586.1
-
3
335
AK308411.1
-
3
333
AM085310.1
-
1
84
AY377974.1
-
3
303
BC018349.1
-
3
327
4338834
BM148203.1
-
3
395
BT009836.1
-
2
135
BU537824.1
-
3
311
DB122945.1
-
3
376
DB148957.1
-
3
403
DC391353.1
-
3
187
JX459579.1
-
3
303
S80876.1
-
1
372
U40462.1
-
3
303
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
mesoderm development
covalent chromatin modification
lymphocyte differentiation
erythrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0