Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM65B SIRNA Assays ›
Gene Symbol:
FAM65B
Gene Name:
family with sequence similarity 65 member B
Gene Aliases:
C6orf32, DFNB104, DIFF40, DIFF48, MYONAP, PL48
Chromosome Location:
Chr.6: 24804281 - 25042288 on Build GRCh38
UniGene ID:
Hs.559459
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43407
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM65B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286445.1
NP_001273374.1
2, 3
341
NM_001286446.1
NP_001273375.1
2, 3
444
NM_001286447.1
NP_001273376.1
2, 3
541
NM_014722.3
NP_055537.2
2, 3
479
NM_015864.3
NP_056948.2
2, 3
541
XM_006715275.2
XP_006715338.1
2, 3
558
XM_006715277.1
XP_006715340.1
2, 3
479
XM_006715279.2
XP_006715342.1
2, 3
536
XM_006715281.3
XP_006715344.1
2, 3
558
XM_011515007.1
XP_011513309.1
3, 4
1067
XM_011515008.2
XP_011513310.1
3, 4
398
XM_011515009.1
XP_011513311.1
2, 3
270
XM_011515010.2
XP_011513312.1
2, 3
311
XM_011515012.1
XP_011513314.1
2, 3
558
XM_017011523.1
XP_016867012.1
2, 3
233
XM_017011524.1
XP_016867013.1
2, 3
487
XM_017011525.1
XP_016867014.1
2, 3
479
XM_017011526.1
XP_016867015.1
2, 3
479
XM_017011527.1
XP_016867016.1
2, 3
311
GenBank mRNA
AB002384.1
-
2, 3
277
AK299047.1
-
2, 3
249
AK300138.1
-
2, 3
489
AK300950.1
-
2, 3
294
BC001232.1
-
2, 3
531
3453890
CN313643.1
-
2, 3
541
DA258515.1
-
2, 3
343
DC406827.1
-
2, 3
445
DC428807.1
-
2, 3
341
U49187.1
-
2, 3
403
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006253930, XM_008771639, XM_008771637, XM_008771638, XM_006253929, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
sensory perception of sound
skeletal muscle fiber development
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0