Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039490.1 | NP_001034579.1 | 3 | 223 | ||
NM_004357.4 | NP_004348.2 | 4 | 289 | |||
NM_139029.1 | NP_620598.1 | 4 | 285 | |||
NM_139030.3 | NP_620599.1 | 3 | 227 | |||
GenBank mRNA | AK130369.1 | - | 4 | 328 | ||
AK223186.1 | - | 4 | 232 | |||
AK293073.1 | - | 4 | 256 | |||
AL161965.1 | - | 4 | 251 | |||
AU099249.1 | - | 4 | 289 | |||
BC001374.2 | - | 3 | 228 | 3051297 | ||
BC013302.2 | - | 4 | 261 | 2958615 | ||
BQ957432.1 | - | 3 | 197 | |||
BT007397.1 | - | 2 | 145 | |||
BT020132.1 | - | 2 | 145 | |||
CR456826.1 | - | 2 | 145 | |||
CR542098.1 | - | 2 | 145 | |||
D29963.1 | - | 3 | 229 | |||
U14650.1 | - | 3 | 204 |