Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53I11 SIRNA Assays ›
Gene Symbol:
TP53I11
Gene Name:
tumor protein p53 inducible protein 11
Gene Aliases:
PIG11
Chromosome Location:
Chr.11: 44932348 - 44951306 on Build GRCh38
UniGene ID:
Hs.554791
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43454
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53I11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258320.1
NP_001245249.1
10
2851
NM_001258321.1
NP_001245250.1
8
2775
NM_001258322.1
NP_001245251.1
8
2705
NM_001258323.2
NP_001245252.1
8
2532
NM_001258324.1
NP_001245253.1
7
2659
NM_001318384.1
NP_001305313.1
8
2540
NM_001318385.1
NP_001305314.1
8
2736
NM_001318386.1
NP_001305315.1
8
2770
NM_001318387.1
NP_001305316.1
5
2301
NM_001318388.1
NP_001305317.1
6
2398
NM_001318389.1
NP_001305318.1
7
2380
NM_001318390.1
NP_001305319.1
7
2652
NM_006034.4
NP_006025.2
7
2457
NR_134612.1
8
2588
XM_005253227.4
XP_005253284.1
8
2552
XM_011520475.2
XP_011518777.1
8
2560
XM_011520476.2
XP_011518778.1
8
2581
XM_011520477.2
XP_011518779.1
8
2604
XM_011520478.2
XP_011518780.1
8
2513
XM_017018580.1
XP_016874069.1
8
2420
XM_017018581.1
XP_016874070.1
7
2254
GenBank mRNA
AF010315.1
-
1
1321
AK026607.1
-
7
2429
AK054815.1
-
8
2512
AK056379.1
-
8
2468
AK096176.1
-
7
2053
BC000443.2
-
1
250
BC003010.1
-
1
250
BC045666.1
-
10
2803
5298525
BC065557.1
-
1
378
Back To Top
More Information
Gene Ontology Categories:
Function(s)
response to stress
negative regulation of cell proliferation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0