Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 3 | 412 | ||
NM_177937.2 | NP_808800.1 | 3 | 404 | |||
GenBank mRNA | AF236056.1 | - | 3 | 353 | ||
AK027717.1 | - | 3 | 375 | |||
AK075542.1 | - | 3 | 346 | |||
AK075557.1 | - | 3 | 316 | |||
AY358593.1 | - | 3 | 317 | |||
BC001740.1 | - | 3 | 327 | 3351316 | ||
BQ216766.1 | - | 371 | ||||
CR457201.1 | - | 2 | 203 | |||
DA116177.1 | - | 3 | 405 | |||
EU794616.1 | - | 3 | 371 |