Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039490.1 | NP_001034579.1 | 7 | 719 | ||
NM_004357.4 | NP_004348.2 | 8 | 785 | |||
NM_139029.1 | NP_620598.1 | 8 | 781 | |||
NM_139030.3 | NP_620599.1 | 7 | 723 | |||
GenBank mRNA | AK130369.1 | - | 7 | 1149 | ||
AK223186.1 | - | 8 | 728 | |||
AK293073.1 | - | 8 | 752 | |||
AL161965.1 | - | 7 | 1072 | |||
BC001374.2 | - | 7 | 724 | 3051297 | ||
BC013302.2 | - | 8 | 757 | 2958615 | ||
BQ957432.1 | - | 7 | 693 | |||
BT007397.1 | - | 6 | 641 | |||
BT020132.1 | - | 6 | 641 | |||
CR456826.1 | - | 6 | 641 | |||
CR542098.1 | - | 6 | 641 | |||
D29963.1 | - | 7 | 725 | |||
U14650.1 | - | 7 | 700 |