Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278618.1 | NP_001265547.1 | 3, 4 | 473 | ||
NM_002827.3 | NP_002818.1 | 4, 5 | 564 | |||
GenBank mRNA | AK290638.1 | - | 4, 5 | 531 | ||
AK299830.1 | - | 3, 4 | 411 | |||
AK316563.1 | - | 4, 5 | 506 | |||
AU117677.1 | - | 4, 5 | 531 | |||
BC015660.2 | - | 4, 5 | 493 | 4844022 | ||
BC018164.1 | - | 4, 5 | 474 | 4338750 | ||
BT006752.1 | - | 4, 5 | 357 | |||
M31724.1 | - | 4, 5 | 447 | |||
M33689.1 | - | 4, 5 | 429 |