Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 7 | 938 | ||
NM_177937.2 | NP_808800.1 | 7 | 930 | |||
GenBank mRNA | AF236056.1 | - | 7 | 879 | ||
AK027717.1 | - | 7 | 901 | |||
AK075542.1 | - | 7 | 872 | |||
AK075557.1 | - | 7 | 842 | |||
AK092943.1 | - | 8 | 818 | |||
AY358593.1 | - | 7 | 843 | |||
BC001740.1 | - | 7 | 853 | 3351316 | ||
CR457201.1 | - | 6 | 729 | |||
EU794616.1 | - | 7 | 897 |